Please login to your online account to display your discounted pricing

Alfa Aesar™ Hafnium(IV) oxide, Spectrographic Grade, 99.9% (metals basis excluding Zr), Zr typically <80ppm

Chemical Identifiers

Chemical Name or Material Hafnium(IV) oxide
CAS 12055-23-1
Purity Grade Spectrographic Grade
Formula Weight 210.49
MDL Number MFCD00003565
Merck Index 14,4588
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1183604

  SDS
alfa aesar™
1183604
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1183609

  SDS
alfa aesar™
1183609
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 210.49
Density 9.68g/mL
Merck Index 14,4588
TSCA TSCA
Quantity 2g
Purity Grade Spectrographic Grade
Chemical Name or Material Hafnium(IV) oxide
Odor Odorless
Assay 99.9%
Melting Point 2774°C
Assay Percent Notes (metals basis excluding Zr), Zr typically <80ppm
Mesh Size −325 Mesh
CAS 12055-23-1
MDL Number MFCD00003565
Physical Form Powder