missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Hafnium(IV) oxide, Spectrographic Grade, 99.9% (metals basis excluding Zr), Zr typically <80ppm

Manufacturer:  Alfa Aesar™ 1183609

 View more versions of this product

Catalog No. AA1183609



Specifications

Specifications

Hafnium(IV) oxide
(metals basis excluding Zr), Zr typically <80ppm
14,4588
Powder
Sprectrographic
−325 Mesh
2774°C
12055-23-1
MFCD00003565
210.49
99.9%
9.68g/mL
Odorless
10g

Chemical Identifiers

12055-23-1
MFCD00003565
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents