Please login to your online account to display your discounted pricing

Hafnium(IV) oxide, 99.99%, (trace metal basis), ACROS Organics™

Chemical Identifiers

Chemical Name or Material Hafnium(IV) oxide
CAS 12055-23-1
Purity Grade Trace Metal Basis
Assay 85% (Hf typical)
Assay Percent Range 99.99%
Molecular Formula HfO2
Linear Formula HfO2
Formula Weight 210.49
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC193140010

  SDS
thermo fisher scientific
AC193140010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC193140050

  SDS
thermo fisher scientific
AC193140050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade Trace Metal Basis
Physical Form Powder
Formula Weight 210.49
Quantity 1g
Color White
Assay 85% (Hf typical)
Density 9.6800g/mL
Chemical Name or Material Hafnium(IV) oxide
Assay Percent Range 99.99%
Molecular Formula HfO2
CAS 12055-23-1
Packaging Glass bottle
Linear Formula HfO2
Total Trace Metal Impurities 100ppm max.

For ultra trace analysis