missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hafnium(IV) oxide, 99.99% (metals basis excluding Zr), Zr <100ppm, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04062006
Description
Hafnium(IV) oxide is used as Intermediates, Paint additives and coating additives, metal Products. And it is also used in optical coatings, as a refractory material in the insulation of such devices as thermocouples.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Hafnium(IV) oxide | |
12055-23-1 | |
5 g | |
HfO2 | |
14,4588 | |
WIHZLLGSGQNAGK-UHFFFAOYSA-N | |
hafnium(4+) dioxidandiide | |
210.49 | |
Odorless |
2774°C | |
Powder | |
(metals basis excluding Zr), Zr <100ppm | |
MFCD00003565 | |
Insoluble in water. | |
[O--].[O--].[Hf+4] | |
210.49 | |
99.99% | |
9.68 g/mL |
Chemical Identifiers
12055-23-1 | |
210.49 | |
WIHZLLGSGQNAGK-UHFFFAOYSA-N | |
[O--].[O--].[Hf+4] |
HfO2 | |
MFCD00003565 | |
hafnium(4+) dioxidandiide |
Safety and Handling
EINECSNumber : 235-013-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only