Please login to your online account to display your discounted pricing

Alfa Aesar™ Hafnium(IV) oxide, 99.95%, (metals basis excluding Zr), Zr typically <0.5%

Chemical Identifiers

Chemical Name or Material Hafnium(IV) oxide
CAS 12055-23-1
Formula Weight 210.49
MDL Number MFCD00003565
Merck Index 14,4588
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3566603

  SDS
alfa aesar™
3566603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3566606

  SDS
alfa aesar™
3566606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3566614

  SDS
alfa aesar™
3566614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 210.49
TSCA TSCA
Quantity 1g
Density 9.68g/mL
Mesh Size −325 Mesh
MDL Number MFCD00003565
Chemical Name or Material Hafnium(IV) oxide
CAS 12055-23-1
Odor Odorless
Assay 99.95%
Physical Form Powder
Melting Point 2774°C
Merck Index 14,4588
Assay Percent Notes (metals basis excluding Zr), Zr typically <0.5%