Please login to your online account to display your discounted pricing

Alfa Aesar™ Hafnium(IV) fluoride, 99.9% (metals basis excluding Zr), Zr 2-4%

Chemical Identifiers

Chemical Name or Material Hafnium(IV) fluoride
CAS 13709-52-9
Formula Weight 254.48
MDL Number MFCD00016127
UN Number UN3290
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3639206

  SDS
alfa aesar™
3639206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3639214

  SDS
alfa aesar™
3639214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 254.48
Chemical Name or Material Hafnium(IV) fluoride
TSCA TSCA
Particle Size 3 to 6mm
Physical Form Sintered Lump
Assay 99.9%
Sensitivity Moisture Sensitive
Assay Percent Notes (metals basis excluding Zr), Zr 2 to 4%
MDL Number MFCD00016127
CAS 13709-52-9
UN Number UN3290
Quantity 5g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H310-H330-H314-H318
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLID, CORROSIVE, INORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H310-H330-H314-H318
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.