Please login to your online account to display your discounted pricing

Hafnium(IV) fluoride 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Hafnium(IV) fluoride
Assay ≥97.5% (ex fluor)
Assay Percent Range 98%
Molecular Formula F4 Hf
Linear Formula HfF4
Formula Weight 254.48
MDL Number MFCD00016127
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC383160250

  SDS
thermo fisher scientific
AC383160250
Plastic bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5% (ex fluor)
Physical Form Powder
Formula Weight 254.48
Quantity 25g
Color Off-white
Linear Formula HfF4
Chemical Name or Material Hafnium(IV) fluoride
Packaging Plastic bottle
Assay Percent Range 98%
Molecular Formula F4 Hf
MDL Number MFCD00016127
CAS 13709-52-9
Zirconium (Zr) 1% max.
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Contact with acids liberates very toxic gas.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.