missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hafnium(IV) chloride, White powder, 99.9% (Metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 4561214

 View more versions of this product

Catalog No. AA4561214



Specifications

Specifications

Hafnium(IV) chloride
99.9% (Metals basis)
MFCD00011034
PDPJQWYGJJBYLF-UHFFFAOYSA-J
tetrachlorohafnium
37715
Powder
Moisture sensitive
25g
13499-05-3
Cl4Hf
hafnium tetrachloride, hafnium iv chloride, hafnium chloride hfcl4 , t-4, hfcl4, hafnium iv chloride,hafnium tetrachloride, hafnium iv chloride, hafnium chloride hfcl4 , t-4, hafnium iv chloride, sublimed grade, hafnium iv chloride, purum at, hafnium iv chloride, purified by sublimation trace metals basis
Cl[Hf](Cl)(Cl)Cl
320.29
320.3
Irritating
White

Chemical Identifiers

13499-05-3
320.29
PDPJQWYGJJBYLF-UHFFFAOYSA-J
37715
Cl[Hf](Cl)(Cl)Cl
Cl4Hf
MFCD00011034
hafnium tetrachloride, hafnium iv chloride, hafnium chloride hfcl4 , t-4, hfcl4, hafnium iv chloride,hafnium tetrachloride, hafnium iv chloride, hafnium chloride hfcl4 , t-4, hafnium iv chloride, sublimed grade, hafnium iv chloride, purum at, hafnium iv chloride, purified by sublimation trace metals basis
tetrachlorohafnium
Safety and Handling

Safety and Handling

Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.

H314-H318: Causes severe skin burns and eye damage. Causes serious eye damage.

DOTInformation : DOT Class: 8, Packing Group: II

EINECSNumber : 236-826-5

SDS Documents