missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hafnium(IV) chloride, 99%, -80 mesh, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC293251000

 View more versions of this product

Catalog No. AC293251000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Hafnium(IV) chloride
13499-05-3
HfCl4
MFCD00011034
hafnium chloride hfcl4 , t-4, hafnium iv chloride, hafnium iv chloride, purified by sublimation trace metals basis, hafnium iv chloride, purum at, hafnium iv chloride, sublimed grade, hafnium iv chloride,hafnium tetrachloride, hafnium tetrachloride, hfcl4
PDPJQWYGJJBYLF-UHFFFAOYSA-J
tetrachlorohafnium
37715
Powder
Glass bottle
White
100g
-80 mesh
98.5% min. (Argentometry)
HfCl4
14, 4588
Solubility in water: reacts. Other solubilities: soluble in methanol,acetone
Cl[Hf](Cl)(Cl)Cl
320.29
320.29
99%
Solubility in water: reacts. Other solubilities: soluble in methanol, acetone
319.0°C

Chemical Identifiers

13499-05-3
320.29
PDPJQWYGJJBYLF-UHFFFAOYSA-J
37715
Cl[Hf](Cl)(Cl)Cl
HfCl4
MFCD00011034
hafnium chloride hfcl4 , t-4, hafnium iv chloride, hafnium iv chloride, purified by sublimation trace metals basis, hafnium iv chloride, purum at, hafnium iv chloride, sublimed grade, hafnium iv chloride,hafnium tetrachloride, hafnium tetrachloride, hfcl4
tetrachlorohafnium
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 236-826-5

SDS Documents