Please login to your online account to display your discounted pricing

Alfa Aesar™ Hafnium iodide, ultra dry, 98.5% (metals basis)

$170.86 - $170.86

Chemical Identifiers

Chemical Name or Material Hafnium iodide
CAS 13777-23-6
Name Note Ultra dry
Formula Weight 686.11
MDL Number MFCD00016128
UN Number UN3260
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3573304

 
alfa aesar™
3573304
2g Each for $170.86

AA3573309

 
alfa aesar™
3573309
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 686.11
Melting Point 400°C (vac.) (subl.)
Packaging Ampule
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Name Note Ultra dry
Physical Form Powder
Assay 98.5%
Sensitivity Moisture Sensitive
CAS 13777-23-6
Chemical Name or Material Hafnium iodide
TSCA TSCA
Assay Percent Notes (metals basis)
MDL Number MFCD00016128
UN Number UN3260
Quantity 2g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Storage Note 1
  • Ampouled under argon
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

  • Ampouled under argon

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

  • Ampouled under argon