missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Hafnium foil, 0.25mm (0.01 in.) thick, annealed, 99.5% (metals basis excluding Zr), Zr nominal 3%

Manufacturer:  Alfa Aesar™ 10793FP

 View more versions of this product

Catalog No. AA10793FP



Specifications

Specifications

Hafnium foil
80 x 250mm
7440-58-6
Hf
and compounds, atom, celtium, foil, hafnio, hafnium, elemental, hsdb 552, powder, unii-x71938l1do, wire
[Hf]
Odorless
23986
Foil
≈13.3g/50x80mm
0.25mm (0.01 in.) thick, Annealed
(metals basis excluding Zr), Zr nominal 3%
MFCD00011032
VBJZVLUMGGDVMO-UHFFFAOYSA-N
hafnium
178.49
CHEBI:33343
99.5%

Chemical Identifiers

7440-58-6
178.49
VBJZVLUMGGDVMO-UHFFFAOYSA-N
23986
hafnium
Hf
MFCD00011032
and compounds, atom, celtium, foil, hafnio, hafnium, elemental, hsdb 552, powder, unii-x71938l1do, wire
CHEBI:33343
[Hf]
Safety and Handling

Safety and Handling

EINECSNumber : 231-166-4

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents