Please login to your online account to display your discounted pricing

Alfa Aesar™ Hafnium dinitrate oxide, 99.9% (metals basis excluding Zr), Zr <0.1%, 10% w/v aq. soln.

Chemical Identifiers

Chemical Name or Material Hafnium dinitrate oxide
Linear Formula HfO(NO3)2
Formula Weight 318.5
MDL Number MFCD00798524
UN Number UN3218
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA44246AB

  SDS
alfa aesar™
44246AB
10mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA44246AD

  SDS
alfa aesar™
44246AD
50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 318.5
Chemical Name or Material Hafnium dinitrate oxide
Physical Form Liquid
Assay 99.9%
Assay Percent Notes (metals basis excluding Zr), Zr <0.1%
Linear Formula HfO(NO3)2
MDL Number MFCD00798524
UN Number UN3218
Quantity 10mL
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H272-H315-H319
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P221-P210-P220-P280-P305+P351+P338-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Keep/Store away from clothing/combustible materials.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Concentration 10% w/v aq. soln.
DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, AQUEOUS SOLUTION, N.O.S .
Safety and Handling

Danger

GHS P Statement
P221-P210-P220-P280-P305+P351+P338-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Keep/Store away from clothing/combustible materials.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H272-H315-H319
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.