missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Hafnium dinitrate oxide, 99.9% (metals basis excluding Zr), Zr <0.1%, 10% w/v aq. soln.

Manufacturer:  Alfa Aesar™ 44246AD

 View more versions of this product

Catalog No. AA44246AD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Hafnium dinitrate oxide
HfO(NO3)2
UN3218
318.5
99.9%
50mL
(metals basis excluding Zr), Zr <0.1%
MFCD00798524
Fully miscible in water.
Liquid
10% w/v aq. soln.

Chemical Identifiers

MFCD00798524
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

DOTInformation : Transport Hazard Class: 5.1; Packaging Group: III; Proper Shipping Name: NITRATES, INORGANIC, AQUEOUS SOLUTION, N.O.S .

TSCA : No

Recommended Storage : Ambient temperatures

Documents