missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hafnium dichloride oxide octahydrate, White powder, >98% (Metals basis excluding Zr), Alfa Aesar™

For Synthesis of Hafnium (IV) Oxide

Manufacturer:  Alfa Aesar 1183314

 View more versions of this product

Catalog No. AA1183314



Specifications

Specifications

Hafnium dichloride oxide octahydrate
>98% (Metals basis excluding Zr)
MFCD00150417
CWGPOJHZSBOIPU-UHFFFAOYSA-L
hafnium;dichloride;nonahydrate
131675691
Powder
White
14456-34-9
Cl2H18HfO9-2
hafnium iv dichloride oxide octahydrate 98+%-hf zr
O.O.O.O.O.O.O.O.O.[Cl-].[Cl-].[Hf]
411.525
409.52 (265.40 Anhydrous)
Pungent
25g

Chemical Identifiers

14456-34-9
411.525
CWGPOJHZSBOIPU-UHFFFAOYSA-L
131675691
O.O.O.O.O.O.O.O.O.[Cl-].[Cl-].[Hf]
Cl2H18HfO9-2
MFCD00150417
hafnium iv dichloride oxide octahydrate 98+%-hf zr
hafnium;dichloride;nonahydrate
Safety and Handling

Safety and Handling

H314-H318-H303: Causes severe skin burns and eye damage. Causes serious eye damage. May be harmful if swallowed.

DOTInformation : DOT Class: 8, Packing Group: III

EINECSNumber : 237-345-3

Documents