Please login to your online account to display your discounted pricing

Alfa Aesar™ Hafnium dichloride oxide octahydrate, Puratronic™, 99.998% (metals basis excluding Zr), Zr <1%

Chemical Identifiers

Chemical Name or Material Hafnium dichloride oxide octahydrate
CAS 14456-34-9
Purity Grade Puratronic™
Linear Formula HfCl2O·8H2O
Formula Weight 409.52 (265.40 Anhydrous)
MDL Number MFCD00150417
UN Number UN3260
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1070503

  SDS
alfa aesar™
1070503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1070506

  SDS
alfa aesar™
1070506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1070514

  SDS
alfa aesar™
1070514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 409.52 (265.40 Anhydrous)
Health Hazard 1 Danger
RTECS Number MG4800000
Physical Form Powder
UN Number UN3260
Assay Percent Notes (metals basis excluding Zr), Zr <1%
Sensitivity Hygroscopic
MDL Number MFCD00150417
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
Linear Formula HfCl2O·8H2O
Quantity 1g
Purity Grade Puratronic™
TSCA TSCA
Assay 99.998%
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Density 2.2g/mL
CAS 14456-34-9
Chemical Name or Material Hafnium dichloride oxide octahydrate
Color White
Odor Pungent
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.