missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Hafnium dichloride oxide octahydrate, Puratronic™, 99.998% (metals basis excluding Zr), Zr <1%

Manufacturer:  Alfa Aesar™ 1070514

 View more versions of this product

Catalog No. AA1070514



Specifications

Specifications

Hafnium dichloride oxide octahydrate
(metals basis excluding Zr), Zr <1%
HfCl2O·8H2O
UN3260
CWGPOJHZSBOIPU-UHFFFAOYSA-L
hafnium;dichloride;nonahydrate
131675691
Powder
Puratronic™
Pungent
White
14456-34-9
Cl2H18HfO9-2
MFCD00150417
hafnium iv dichloride oxide octahydrate 98+%-hf zr
O.O.O.O.O.O.O.O.O.[Cl-].[Cl-].[Hf]
411.525
409.52 (265.40 Anhydrous)
99.998%
2.2g/mL
Hygroscopic
25g

Chemical Identifiers

14456-34-9
411.525
CWGPOJHZSBOIPU-UHFFFAOYSA-L
131675691
O.O.O.O.O.O.O.O.O.[Cl-].[Cl-].[Hf]
Cl2H18HfO9-2
MFCD00150417
hafnium iv dichloride oxide octahydrate 98+%-hf zr
hafnium;dichloride;nonahydrate
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

RTECSNumber : MG4800000

TSCA : TSCA

Documents