missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Hafnium crystal bar milled chips, 99.7% (metals basis excluding Zr), Zr nominal 3%

Manufacturer:  Alfa Aesar™ 1020414

 View more versions of this product

Catalog No. AA1020414



Specifications

Specifications

Hafnium
7440-58-6
Hf
and compounds
[Hf]
Odorless
23986
Crystal Bar Milled Chips
25g
(metals basis excluding Zr), Zr nominal 3%
MFCD00011032
VBJZVLUMGGDVMO-UHFFFAOYSA-N
hafnium
178.49
CHEBI:33343
99.7%

Chemical Identifiers

7440-58-6
178.49
VBJZVLUMGGDVMO-UHFFFAOYSA-N
23986
hafnium
Hf
MFCD00011032
and compounds
CHEBI:33343
[Hf]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents