missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hafnium carbide, Gray powder, 99.5% (Metals basis excluding Zr), Alfa Aesar™

Manufacturer:  Alfa Aesar 1256703

 View more versions of this product

Catalog No. AA1256703



Specifications

Specifications

Hafnium carbide
99.5% (Metals basis excluding Zr)
MFCD00016126
Powder
Gray
12069-85-1
CHf
190.54
Odorless
1g

Chemical Identifiers

12069-85-1
CHf
MFCD00016126
Safety and Handling

Safety and Handling

Take precautionary measures against static discharges.

H228: Flammable solid.

DOTInformation : DOT Class: 4.1, Packing Group: III

EINECSNumber : 235-114-1

Documents