Please login to your online account to display your discounted pricing

Hafnium carbide, Gray powder, 99.5% (Metals basis excluding Zr), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Hafnium carbide
CAS 12069-85-1
Physical Form Powder
Assay Percent Range 99.5% (Metals basis excluding Zr)
Molecular Formula CHf
Formula Weight 190.54
MDL Number MFCD00016126
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1256703

  SDS
alfa aesar
1256703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1256709

  SDS
alfa aesar
1256709
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1256718

  SDS
alfa aesar
1256718
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 190.54
Quantity 1g
Color Gray
DOT Information DOT Class: 4.1, Packing Group: III
Chemical Name or Material Hafnium carbide
EINECS Number 235-114-1
Odor Odorless
Assay Percent Range 99.5% (Metals basis excluding Zr)
Molecular Formula CHf
MDL Number MFCD00016126
CAS 12069-85-1
Health Hazard 1 H228: Flammable solid.
Health Hazard 2 Take precautionary measures against static discharges.
Safety and Handling

H228: Flammable solid.

Take precautionary measures against static discharges.