missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Hafnium 1,1,1-trifluoro-2,4-pentanedionate

Manufacturer:  Alfa Aesar™ 1927502

 View more versions of this product

Catalog No. AA1927502



Specifications

Specifications

Hafnium 1,1,1-trifluoro-2,4-pentanedionate
0.5g
MFCD00078027
Soluble in acetone, cyclohexane
Powder
128°C to 129°C
17475-68-2
Hafnium trifluoroacetylacetonate
790.81

Chemical Identifiers

17475-68-2
MFCD00078027
Hafnium trifluoroacetylacetonate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents