Please login to your online account to display your discounted pricing

Alfa Aesar™ Hafnium 1,1,1-trifluoro-2,4-pentanedionate

$226.01 - $226.01

Chemical Identifiers

Chemical Name or Material Hafnium 1,1,1-trifluoro-2,4-pentanedionate
CAS 17475-68-2
Formula Weight 790.81
MDL Number MFCD00078027
Synonym Hafnium trifluoroacetylacetonate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1927502

  SDS
alfa aesar™
1927502
0.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1927504

  SDS
alfa aesar™
1927504
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1927509

  SDS
alfa aesar™
1927509
10g Each for $226.01
Description & Specifications

Specifications

Formula Weight 790.81
Chemical Name or Material Hafnium 1,1,1-trifluoro-2,4-pentanedionate
Physical Form Powder
MDL Number MFCD00078027
CAS 17475-68-2
Synonym Hafnium trifluoroacetylacetonate
Solubility Information Soluble in acetone, cyclohexane
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 128° to 129°C
Quantity 0.5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.