Please login to your online account to display your discounted pricing

HA14-1, Tocris

Bcl-2 inhibitor; Induces apoptosis

$181.19 - $762.58

Chemical Identifiers

Chemical Name or Material HA14-1
Assay Percent Range >98%
Molecular Formula C17H17BrN2O5
Formula Weight 409.23
Synonym 2-Amino-6-bromo-α-cyano-3-(ethoxycarbonyl)-4H-1-benzopyran-4-acetic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

154110

 
r & d systems
RDS154110
10mg Each for $181.19

154150

 
r & d systems
RDS154150
50mg Each for $762.58
Description & Specifications

Specifications

Formula Weight 409.23
Quantity 10mg
CAS 65673-63-4
Chemical Name or Material HA14-1
Assay Percent Range >98%
Molecular Formula C17H17BrN2O5
Synonym 2-Amino-6-bromo-α-cyano-3-(ethoxycarbonyl)-4H-1-benzopyran-4-acetic acid ethyl ester
Recommended Storage Desiccate at -20°C
Safety and Handling

Desiccate at -20°C