missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ HA (Hemagglutinin) Ab-1, Rabbit Polyclonal Antibody

Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ HA (Hemagglutinin) Ab-1, Rabbit Polyclonal Antibody.

$254.18 - $1,266.27

Specifications

Antigen HA (Hemagglutinin) Ab-1
Concentration 1mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
RB1438P Thermo Scientific™ Lab Vision™
RB1438P
Purified Ab with BSA and Azide 1mL Each for $1,165.41
RB1438P0 Thermo Scientific™ Lab Vision™
RB1438P0
Purified Ab with BSA and Azide 100μL Each for $254.18
RB1438P1 Thermo Scientific™ Lab Vision™
RB1438P1
Purified Ab with BSA and Azide 500μL Each for $742.12
RB1438P1ABX Thermo Scientific™ Lab Vision™
RB1438P1ABX
Ab Without BSA/Azide (Frozen) 500μL Each for $813.94
RB1438PABX Thermo Scientific™ Lab Vision™
RB1438PABX
Ab Without BSA/Azide (Frozen) 1mL Each for $1,266.27
Specifications

Description

Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ HA (Hemagglutinin) Ab-1, Rabbit Polyclonal Antibody.

A small segment of the viral hemagglutinin coat protein is widely used as an epitope tag to facilitate purification and functional analysis of proteins of interest. Epitope tags are short peptide sequences that are easily recognized by readily available tag-specific antibodies. Ligation of epitope-tag encoding DNA to cloned DNAs of interest produces epitope tagged fusion proteins that can then be subjected to antibody-dependent experimental procedures. Epitope tagging techniques are widely used in the purification, identification, and functional analysis of proteins. Expression vectors for producing tag fusion proteins in a variety of organisms (including bacteria, yeast, insect, and mammalian cells) are commercially available. Some of the more commonly used fusion tags include: c-myc, FLAG, GFP, GST, HA, His, amd MBP.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Proteins displaying the YPYDVPDYA HA-tag.

Epitope: aa YPYDVPDYA (HA-tag)

Immunogen: Synthetic peptide of sequence CYPYDVPDYASL

Positive Control: Proteins displaying the YPYDVPDYA HA-tag

Cellular Localization: Depends upon the localization of the parent protein tagged with the HA tag

Recommended for:

  • Western Blotting
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

HA (Hemagglutinin) Ab-1
Western Blot
Unlabeled
Rabbit
Epitope Tags
Synthetic peptide of sequence CYPYDVPDYASL
1mg/mL
Polyclonal
Concentrated
RUO
Primary
IgG