Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ HA (Hemagglutinin) Ab-1, Rabbit Polyclonal Antibody


Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ HA (Hemagglutinin) Ab-1, Rabbit Polyclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  RB1438P1ABX

 View more versions of this product

Catalog No. RB1438P1ABX

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity All
Antigen HA (Hemagglutinin) Ab-1
Isotype IgG
Host Species Rabbit
Applications Western Blotting
Format Concentrated
Immunogen Synthetic peptide of sequence CYPYDVPDYASL
Quantity 500µL
Regulatory Status RUO
Formulation Ab Without BSA/Azide (Frozen)
Conjugate Unlabeled
Concentration 1mg/mL
Type Primary
Research Discipline Epitope Tags
Classification Polyclonal

Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ HA (Hemagglutinin) Ab-1, Rabbit Polyclonal Antibody.

A small segment of the viral hemagglutinin coat protein is widely used as an epitope tag to facilitate purification and functional analysis of proteins of interest. Epitope tags are short peptide sequences that are easily recognized by readily available tag-specific antibodies. Ligation of epitope-tag encoding DNA to cloned DNAs of interest produces epitope tagged fusion proteins that can then be subjected to antibody-dependent experimental procedures. Epitope tagging techniques are widely used in the purification, identification, and functional analysis of proteins. Expression vectors for producing tag fusion proteins in a variety of organisms (including bacteria, yeast, insect, and mammalian cells) are commercially available. Some of the more commonly used fusion tags include: c-myc, FLAG, GFP, GST, HA, His, amd MBP.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Proteins displaying the YPYDVPDYA HA-tag.

Epitope: aa YPYDVPDYA (HA-tag)

Immunogen: Synthetic peptide of sequence CYPYDVPDYASL

Positive Control: Proteins displaying the YPYDVPDYA HA-tag

Cellular Localization: Depends upon the localization of the parent protein tagged with the HA tag

Recommended for:

  • Western Blotting
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO