missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

HA 130, Tocris™

Selective autotaxin inhibitor

$250.58 - $1,042.86

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C24H19BFNO5S
Formula Weight 463.29
Synonym B-[3-[[4-[[3-[(4-Fluorophenyl)methyl]-2, 4-dioxo-5-thiazolidinylidene]methyl]phenoxy]methyl]phenyl]-boronic acid
Chemical Name or Material HA 130
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
419610 R & D Systems
RDS419610
10mg Each for $250.58
419650 R & D Systems
RDS419650
50mg Each for $1,042.86
Specifications Description & Specifications

Chemical Identifiers

>98%
463.29
HA 130
C24H19BFNO5S
B-[3-[[4-[[3-[(4-Fluorophenyl)methyl]-2, 4-dioxo-5-thiazolidinylidene]methyl]phenoxy]methyl]phenyl]-boronic acid

Specifications

10mg
>98%
463.29
HA 130
1229652-21-4
C24H19BFNO5S
B-[3-[[4-[[3-[(4-Fluorophenyl)methyl]-2, 4-dioxo-5-thiazolidinylidene]methyl]phenoxy]methyl]phenyl]-boronic acid

Safety and Handling

Recommended Storage : Store at -20°C