Please login to your online account to display your discounted pricing

H2L5186303, Tocris™

Potent and selective LPA2 receptor antagonist

$236.06 - $976.15

Chemical Identifiers

Chemical Name or Material H2L5186303
Assay Percent Range >98%
Molecular Formula C26H20N2O8
Formula Weight 488.45
Synonym (Z,Z)-4,4'-[1,3-Phenylenebis(oxy-4,1-phenyleneimino)]bis[4-oxo-2-butenoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

487810

 
r & d systems
RDS487810
10mg Each for $236.06

487850

 
r & d systems
RDS487850
50mg Each for $976.15
Description & Specifications

Specifications

Formula Weight 488.45
Quantity 10mg
Synonym (Z,Z)-4,4'-[1,3-Phenylenebis(oxy-4,1-phenyleneimino)]bis[4-oxo-2-butenoic acid
Chemical Name or Material H2L5186303
Assay Percent Range >98%
CAS 139262-76-3
Molecular Formula C26H20N2O8
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C