Please login to your online account to display your discounted pricing

H-B Instrument™ Durac™ Specific Gravity/Relative Density (g/cm&superscript_3;) Plain Form Hydrometers: for Liquids Lighter Than Water


Available in a variety of models measuring specific gravity/relative density (g/cm&superscript_3;) of liquids lighter than water in various ranges from 0.690 to 3.000

Manufacturer: hb instrument  B618014600RMD

 View more versions of this product

Catalog No. 13-203-110

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) For Liquids Lighter Than Water
Specific Gravity Range 0.800 to 0.910
Recommended Storage Plain form hydrometers should be stored in a clean, dry, safe place. They should not be subjected to extreme temperatures.
Accuracy ±0.001
Increments 0.001
Certifications/Compliance Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST. Hand inspected by U.S. quality team in exclusive triple accredited/registered ISO/IEC 17025:2005 and A2LA Accredited Calibration Laboratory (Cert 2448.01); internationally recognized and accepted by the signatory economies of the ILAC/MRA and located in ISO 14001:2004 Environmental Management System registered facility.
Length (Metric) 330mm