H-B™ Enviro-Safe™ Environmentally Friendly Liquid-In-Glass Pocket Thermometers
Environmentally friendly portable thermometers
Manufacturer:  Hb Instrument B605700500
 View more versions of this product
Catalog No. 13201501
Specifications
Description
Thermometer, Pocket; H-B Enviro-Safe; Liquid-In-Glass; Closed plastic case; Temp. Range: -10deg. to +110deg.C; Total immersion; Non-toxic liquid fill; Length: 160mm
EnviroKleen™ certified thermometers feature green, nontoxic, biodegradable, Enviro-Safe liquid against lead-free, white back glass Total immersion pocket thermometers are a compact 160mm (6.3 in.) long Come with a case to protect from breakage Case options include: Plastic Window - encased in plastic with an open window Metal Window - encased in metal with an open window Closed Plastic - housed in removable plastic case Closed Metal - housed in a removable metal case Aluminum Duplex - encased in nickel-plated brass with an open window and housed in a removable aluminum case Nickel-Plated Brass Window - encased in nickel-plated brass with an open window
Description & Specifications
Specifications
±1°C
Ideal for field studies, waste water treatment and any “on-the-go” applications.
Closed plastic case
160mm
Not Applicable
Manufactured and tested by USA quality team and include a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.
Total
6.3 in.
1°C
-10° to +110°C