HB Instruments H-B™ Enviro-Safe™ General Purpose Liquid-In-Glass Thermometers, Partial Immersion
Certified environmentally safe thermometers replace dangerous mercury
$25.38 - $25.38
Specifications
Certifications/Compliance Individually serialized thermometers are traceable to NIST; include a multi-language Statement of Accuracy and Traceability.
Immersion Type Partial
Length (English) Overall 7.9 in.
Length (Metric) Overall 200mm
For Use With (Application) Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Accuracy Subdivision Temperature Range (English) Temperature Range (Metric) Price Quantity  
Catalog Number Mfr. No. Accuracy Subdivision Temperature Range (English) Temperature Range (Metric) Price Quantity  
13-202-554 Fisher Scientific
B605010000RMD
±1°C 1°C Not Applicable -10° to +110°C Each for $25.38
Specifications
Description
  • Earth friendly, EnviroKleen™ certified thermometers
  • Feature green, non-toxic, biodegradable, Enviro-Safe™ liquid against lead-free, white back glass
  • Meet government, education, and industry mandates to purchase “Green” products
 Applications:
Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
Order Info
Should you require an individualized Thermometer Calibration Report, factory calibrated thermometers or custom calibration services are available.
Certifications
Individually serialized thermometers are traceable to NIST; include a multi-language Statement of Accuracy and Traceability.
Compliance
Manufactured and tested by USA quality team.
Description & Specifications
Specifications
Individually serialized thermometers are traceable to NIST; include a multi-language Statement of Accuracy and Traceability.
7.9 in.
Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
Partial
200mm
50mm (2 in.)