Please login to your online account to display your discounted pricing
H-B Instrument™ Durac™ Waterproof Electronic Thermometer
For reading temperatures in sample reagents, water baths, semi-solids and gases
$100.62 - $100.62
Specifications
Material Stainless steel
View More Specs
Products
Catalog Number Mfr. No. Accuracy Length (English) Length (Metric) Temperature Range (Metric) Price Quantity    
13-201-768
fisher scientific
B609001600
4°C 5 in. 127mm -40° to +232°C Each for $100.62
Description & Specifications
Specifications
Material Stainless steel
  • Feature stainless-steel temperature probe
  • Probe cover with pocket clip included
  • 0.1 resolution
  • Ideal for field studies, cuvettes, test tubes, flasks and beakers
  • Supplied with instructions in English, French, German, Italian, Portuguese and Spanish languauges and Statement of Traceability
  • Tested and calibrated in triple accredited/registered ISO™/IEC 17025:2005, A2LA accredited laboratory against equipment with calibration traceable to NIST™
CE marked, NIST traceable
RoHS compliant