Please login to your online account to display your discounted pricing
H-B Instrument™ Durac™ Water Resistant Electronic Thermometers
Ensure fast and accurate temperature measurement for wet lab applications, wash down areas, food testing and tanks
$82.30 - $82.30
Specifications
For Use With Wet labs, wash down areas, food testing and tanks
Material Stainless steel
View More Specs
Products
Catalog Number Mfr. No. Length (Metric) Probe Product Type Temperature Range (English) Temperature Range (Metric) Price Quantity    
13-201-774
hb instrument
B609001700
12cm Electronic Thermometer -58° to +392°F -50° to +200°C Each for $82.30
Description & Specifications
Specifications
For Use With Wet labs, wash down areas, food testing and tanks
Material Stainless steel
  • Feature stainless-steel temperature probe
  • Include probe cover with pocket clip
  • Maximum/minimum memory feature with 0.1 resolution
  • IP65 rating
  • Supplied with instructions in English, French, German, Italian, Portuguese and Spanish languauges and Statement of Traceability
  • Tested and calibrated in triple accredited/registered ISO™/IEC 17025:2005, A2LA accredited laboratory against equipment with calibration traceable to NIST™ and DKD
CE marked, NIST traceable
RoHS compliant