Please login to your online account to display your discounted pricing

H-B Instrument™ Durac™ Specific Gravity ASTM Hydrometers: for Light Liquids


Available in a variety of models measuring specific gravity of heavy and light liquids in various ranges from 0.650 to 2.000

Manufacturer: fisher scientific  B618910300

 View more versions of this product

Catalog No. 13-202-336

  • / Each
 

Description & Specifications

Specifications

Increments 0.0005
Recommended Storage ASTM hydrometers should be stored in a clean, dry, safe place. They should not be subjected to extreme temperatures.
Certifications/Compliance Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST. Hand inspected by U.S. quality team in exclusive triple accredited/registered ISO/IEC 17025:2005 and A2LA Accredited Calibration Laboratory; internationally recognized and accepted by the signatory economies of the ILAC/MRA and located in ISO 14001:2004 Environmental Management System registered facility. Conforms to American Society of Testing Materials (ASTM) E100 standards.
Accuracy ±0.0005
Length (Metric) 330mm
For Use With (Application) Commonly used in industries such as food and beverage, chemical, pharmacology, transportation, refrigeration and more.
ASTM Number 85H
Specific Gravity Range 0.800 to 0.850
Accessories

ASTM hydrometers should be stored in a clean, dry, safe place. They should not be subjected to extreme temperatures.