Please login to your online account to display your discounted pricing

H-B Instrument™ Durac™ Specific Gravity ASTM Hydrometers: for Light Liquids


Available in a variety of models measuring specific gravity of heavy and light liquids in various ranges from 0.650 to 2.000

Manufacturer: fisher scientific  B618911200

 View more versions of this product

Catalog No. 13-203-106

  • / Each
  • / Case of 10 Each
 

Description & Specifications

Specifications

Recommended Storage ASTM hydrometers should be stored in a clean, dry, safe place. They should not be subjected to extreme temperatures.
For Use With (Application) Commonly used in industries such as food and beverage, chemical, pharmacology, transportation, refrigeration and more.
Specific Gravity Range 0.850 to 0.900
ASTM Number 106H
Accuracy ±0.001
Increments 0.001
Certifications/Compliance Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST. Hand inspected by U.S. quality team in exclusive triple accredited/registered ISO/IEC 17025:2005 and A2LA Accredited Calibration Laboratory; internationally recognized and accepted by the signatory economies of the ILAC/MRA and located in ISO 14001:2004 Environmental Management System registered facility. Conforms to American Society of Testing Materials (ASTM) E100 standards.
Length (Metric) 260mm