Please login to your online account to display your discounted pricing

H-B Instrument™ Durac™ Specific Gravity ASTM Hydrometers: for Heavy Liquids


Available in a variety of models measuring specific gravity of heavy and light liquids in various ranges from 0.650 to 2.000

Manufacturer: hb instrument  B618911500

 View more versions of this product

Catalog No. 13-202-440

  • / Each
 

Description & Specifications

Specifications

Increments 0.0005
Product Type For Heavy Liquids
Recommended Storage ASTM hydrometers should be stored in a clean, dry, safe place. They should not be subjected to extreme temperatures.
Specific Gravity Range 1.000 to 1.050
Certifications/Compliance Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST. Hand inspected by U.S. quality team in exclusive triple accredited/registered ISO/IEC 17025:2005 and A2LA Accredited Calibration Laboratory; internationally recognized and accepted by the signatory economies of the ILAC/MRA and located in ISO 14001:2004 Environmental Management System registered facility. Conforms to American Society of Testing Materials (ASTM) E100 standards.
ASTM Number 111H
Accuracy ±0.0005
For Use With (Application) Commonly used in industries such as food and beverage, chemical, pharmacology, transportation, refrigeration and more.
Length (Metric) 330mm
Accessories

ASTM hydrometers should be stored in a clean, dry, safe place. They should not be subjected to extreme temperatures.