H-B Instrument™ Durac™ ASTM Precision Specific Gravity Hydrometers with Individual Calibration Report: for Heavy Liquids
Available in a variety of individually calibrated models measuring specific gravity of heavy liquids in various ranges from 0.650 to 1.500
Manufacturer:  Hb Instrument B618981900
 View more versions of this product
Catalog No. 13-203-058
Specifications Description & Specifications
Specifications
1.105, 1.125, 1.145
Calibrated and hand inspected by U.S. quality team in exclusive triple accredited/registered ISO/IEC 17025:2005 and A2LA Accredited Calibration Laboratory; internationally recognized and accepted by the signatory economies of the ILAC/MRA and located in ISO 14001:2004 Environmental Management System registered facility. Conforms with American Society of Testing Materials (ASTM) E100 standards.
For Heavy Liquids.
±0.0005
113H
0.0005
1.100 to 1.150
330mm