Please login to your online account to display your discounted pricing

H-B™ Durac™ Specific Gravity/Relative Density Plastic Plain Form Hydrometers


Shatterproof polycarbonate prevents contamination

Manufacturer: fisher scientific  B618000000

 View more versions of this product

 Alert

Polycarbonate hydrometers are not recommended for use with petroleum-based liquids or other corrosives that would potentially affect the plastic material. They should be stored in a clean, dry, safe place and should not be subjected to extreme temperatures.

Catalog No. 13-201-450

  • / Each
 

Description & Specifications

Specifications

Length (Metric) 300mm
For Use With (Application) Ideal in situations where rough handling or contamination could be an issue, such as in the food or beverage industries.
Certifications/Compliance Hydrometers are made and hand inspected by U.S. quality team in exclusive triple accredited/registered ISO/IEC 17025:2005 and A2LA Accredited Calibration Laboratory; internationally recognized and accepted by the signatory economies of the ILAC /MRA and located in ISO 14001:2004 Environmental Management System registered facility. They are packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST.
Increments 0.002
Specific Gravity Range 1.000 to 1.180
Accuracy ±0.002