Please login to your online account to display your discounted pricing

H-B™ Durac™ Baume Plastic Plain Form Hydrometers


Safety and quality protection in one instrument!

Manufacturer: hb instrument  B618000700

 View more versions of this product

 Alert

Polycarbonate hydrometers are not recommended for use with petroleum-based liquids or other corrosives that would potentially affect the plastic material. They should be stored in a clean, dry, safe place and should not be subjected to extreme temperatures.

Catalog No. 13-202-365

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Ideal in situations where rough handling or contamination could be an issue, such as in the food or beverage industries.
Specific Gravity Range 0° to 12°
Accuracy ±0.2deg.
Increments 0.2deg.
Certifications/Compliance Hydrometers are made and hand inspected by U.S. quality team in exclusive triple accredited/registered ISO/IEC 17025:2005 and A2LA Accredited Calibration Laboratory; internationally recognized and accepted by the signatory economies of the ILAC /MRA and located in ISO 14001:2004 Environmental Management System registered facility. Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST.
Length (Metric) 255mm