Please login to your online account to display your discounted pricing

H-B™ Durac™ Baume Plastic Plain Form Hydrometers


Safety and quality protection in one instrument!

Manufacturer: hb instrument  B618000800

 View more versions of this product

 Alert

Polycarbonate hydrometers are not recommended for use with petroleum-based liquids or other corrosives that would potentially affect the plastic material. They should be stored in a clean, dry, safe place and should not be subjected to extreme temperatures.

Catalog No. 13-202-366

  • / Each
 

Description & Specifications

Specifications

Length (Metric) 255mm
Increments 0.2deg.
Accuracy ±0.2deg.
Specific Gravity Range 9° to 21°
Certifications/Compliance Hydrometers are made and hand inspected by U.S. quality team in exclusive triple accredited/registered ISO/IEC 17025:2005 and A2LA Accredited Calibration Laboratory; internationally recognized and accepted by the signatory economies of the ILAC /MRA and located in ISO 14001:2004 Environmental Management System registered facility. Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST.
For Use With (Application) Ideal in situations where rough handling or contamination could be an issue, such as in the food or beverage industries.