missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

H-B Instrument™ Durac™ Plus™ ASTM-Like Partial Immersion Laboratory Thermometers

Safer than their mercury counterparts

Manufacturer:  Hb Instrument B607502100

 View more versions of this product

Catalog No. 13-201-720


 

Specifications Description & Specifications

Specifications

0.6°C
ASTM
100mm
15.5 in.
White Back Glass
-2° to +52°C
37C
Quality control laboratories or where ISO 9000 standards are in place
Partial
395mm
0.2
Blue Organic
Promotions

Promotions