H-B Instrument™ Durac™ Electronic Thermometers
Fast and accurate temperature measurement with maximum/minimum memory
$82.30 - $82.30
Specifications
Certifications/Compliance ISO/IEC 17025:2005, A2LA, NIST, DAkkS
Accuracy 2°C
Includes Flip out stand, multilanguage instructions and statement of traceability, one AAA battery
Resolution 0.1°C
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Dimensions (L x W x H) Length (English) Cable Length (English) Probe Temperature Range (English) Price Quantity  
Catalog Number Mfr. No. Dimensions (L x W x H) Length (English) Cable Length (English) Probe Temperature Range (English) Price Quantity  
13-201-760 Hb Instrument
B609000400
4.9 x 2.5 x 3.7 in. (125 x 6.5 x 9.4cm) 59.05 in. 4.9 in. -14° to +122°F Zone 1, -58° to +158°F Zone 2 Each for $82.30
Specifications
Description
  • Feature flip out stand
  • Data hold with 0.1 resolution
  • Supplied with instructions in English, French, German, Italian, Portuguese and Spanish languauges and Statement of Traceability
  • Tested and calibrated in triple accredited/registered ISO™/IEC 17025:2005, A2LA accredited laboratory against equipment with calibration traceable to NIST™ and DKD
Certifications
CE marked
Compliance
RoHS compliant
Description & Specifications
Specifications
ISO/IEC 17025:2005, A2LA, NIST, DAkkS
2°C
Flip out stand, multilanguage instructions and statement of traceability, one AAA battery
0.1°C