missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
H-B Instrument™ Durac™ Calcium Chloride Salometers
Two models: one measures percent saturation of CaCl2 in the range of 0 to 120%, and the other measures specific gravity/relative density (g/cm3) from 1.000 to 1.280.
Description
- 300mm (12 in.) long
- Lead-free ballast and glass
- Individually serialized
- Temperature standardization: 15°C (60°F)
Applications:
Used in various industries including food, fishing, and transportation.
Storage:
Calcium Chloride Salometers should be stored in a clean, dry, safe place, and should not be subjected to extreme temperatures.
Order Info
Should you require an individual Thermometer Calibration Report, factory calibrated thermometers or custom calibration services are available.
Certifications
Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST. Hand inspected by U.S. quality team.
Specifications
±0.002 Sp. Gr. | |
Traceable to NIST | |
Calcium Chloride Salometers should be stored in a clean, dry, safe place, and should not be subjected to extreme temperatures. | |
1.00/1.28 |
0.002 Specific Gravity | |
300 mm | |
Used in various industries including food, fishing, and transportation. |