missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

H-B Instrument™ Durac™ Bi-Metallic Thermometers: 44mm Dial

With 2.5% or 2° accuracy—ideal for measuring temperatures in pipes, griddles and hot plates

Manufacturer:  H-B Instrument™ B613104400

 View more versions of this product

Catalog No. 13-201-434


 

Specifications Description & Specifications

Specifications

1%
1.75 in.
1.7 in. (44mm)
50mm in liquids or 100mm in gas
7.9 in.
200mm
Polycarbonate
2°C
0° to 100°C
ISO/IEC 17025:2005, A2LA, NIST, DKD
44mm
Petrochemical, oil and gas, waste water, pharmaceutical, compost, food and beverage storage and processing, asphalt, mining, utilities, automotive, marine, photographic
Stainless Steel probe, multilanguage statement of test and calibration, beaker clip with built in adjustment tool
7.9 in.
200mm
Metal
Not Applicable
Promotions

Promotions