missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

H-B Instrument™ Durac™ Bi-Metallic Dial Thermometers - 75mm Dial

Thermometers with NPT connection for measuring temperature in gas and liquid phase

Manufacturer:  H-B Instrument™ B613108200RMD

 View more versions of this product

Catalog No. 13-201-821


 

Specifications Description & Specifications

Specifications

NIST
75mm
Petrochemical, oil and gas, waste water, pharmaceutical, compost, food and beverage storage and processing, asphalt, mining, utilities, automotive, marine, photographic
Stainless Steel probe, Multilanguage Statement of Test and Calibration, 0.5 in. NPT threaded connection
8.8 in.
225mm
Metal
Not Applicable
3 in.
3 in. (75mm)
50mm in liquids or 100mm in gas
8.9 in.
225mm
Polycarbonate
1°C, 2°F
-20° to +120°C
Promotions

Promotions