missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

H-B™ Instruments Double-Safe™ PFA Safety Coated, Liquid-In-Glass Thermometers: Total Immersion

Protect your workplace and ensure a safe environment

Manufacturer:  Fisher Scientific B604001600RMD

 View more versions of this product

Catalog No. 13-202-524


 

Specifications

Description

Thermometer, Total immersion; H-B Instrument; Double-Safe; Enviro-Safe fill; PFA coating; Yellow back glass; Range: -20deg. to +110deg.C ; Div: 1deg.C; Acc: +/-1deg.C; L: 300mm

  • Environmentally safe thermometers are PFA safety coated to keep liquid and glass contained in the event of breakage
  • Lead-free, yellow back glass and black liquid provides easy readability and fewer reading errors
  • Contains non-toxic, biodegradable, Enviro-Safe™ liquid
  • Manufactured and tested by our USA quality team

Applications:

Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.

Order Info

Should you require an individual Thermometer Calibration Report, factory calibrated thermometers or custom calibration services are available.

Certifications

Traceable to NIST™. Include a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.

Description & Specifications

Specifications

±1°C
Chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical industry, medical industry, manufacturers, dairies, breweries and distilleries
Total
300mm
Yellow Back Glass
Not Applicable
Enviro-Safe™
Traceable to NIST. Include a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.
Precision
12 in.
300mm
1°C
-20° to +110°C
Promotions

Promotions