missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

H-B™ Instruments Double-Safe™ PFA Safety Coated, Liquid-In-Glass Thermometers: Partial Immersion

Protect your workplace and ensure a safe environment

Manufacturer:  Fisher Scientific B604000900RMD

 View more versions of this product

Catalog No. 13-202-518


 

Specifications

Description

Thermometer, Partial immersion; H-B Instrument; Double-Safe; Enviro-Safe fill; PFA Coating; Yellow back glass; Range: 0deg. to 230deg.F, Div: 2deg.F, Accry.: +/-2deg.F; L: 300mm (12 in.); Immersion Depth: 76mm

  • Environmentally safe thermometers are PFA safety coated to keep liquid and glass contained in the event of breakage
  • Lead-free, yellow back glass and black liquid provides easy readability and fewer reading errors
  • Contains non-toxic, biodegradable, Enviro-Safe™ liquid
  • Manufactured and tested by our USA quality team

Applications:

Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.

Order Info

Should you require an individual Thermometer Calibration Report, factory calibrated thermometers or custom calibration services are available.

Certifications

Traceable to NIST™. Includes a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.

Description & Specifications

Specifications

±2°F
Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
76mm (3 in.)
Multilanguage instructions, statement of accuracy
300mm
Yellow Back Glass
0° to 230°F
Enviro-Safe™
Traceable to NIST. Include a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.
Precision
Partial
12 in.
300mm
2°F
Not Applicable
Promotions

Promotions