Please login to your online account to display your discounted pricing

H-9 dihydrochloride, Tocris


Protein kinase inhibitor

Quantity 50mg

Manufacturer: r & d systems  RDS039650

 View more versions of this product

Catalog No. 039650

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 324.22
Assay Percent Range >99%
CAS 116700-36-8
Molecular Formula C11H13N3O2S·2HCl
Synonym N-(2-Aminoethyl)-5-isoquinolinesulfonamide dihydrochloride
Chemical Name or Material H-9 dihydrochloride
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material H-9 dihydrochloride
Assay Percent Range >99%
Molecular Formula C11H13N3O2S·2HCl
Formula Weight 324.22
Synonym N-(2-Aminoethyl)-5-isoquinolinesulfonamide dihydrochloride
Accessories

Store at Room Temperature