Gyro Studies Lab Stool Kit
Unique lab stool has a turntable built into the seat.
$610.20 - $610.20
Specifications
For Use With (Application) For gyro studies
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Product Type Price Quantity  
Catalog Number Mfr. No. Includes Product Type Price Quantity  
S68110 Fisher Science Education
18730
Two accelerometers, one rotary mounting arm and two bag toss masses Gyro Studies Kit Each for $610.20
Specifications
Description
LabStool
  • Made from interlocking parts, stool can be disassembled for easy storage
  • Includes glue for permanent assembly
  • Seat is slotted to accept rotational accessories; included insert allows user to develop personalized attachments
  • Gyro Studies Kit
    • Lab stool with bicycle wheel gyroscope, two accelerometers, one rotary mounting arm and two bag toss masses
Description & Specifications
Specifications
For gyro studies