Please login to your online account to display your discounted pricing

GYKI 47261 dihydrochloride, Tocris

Non-competitive AMPA antagonist

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material GYKI 47261 dihydrochloride
Assay Percent Range >99%
Molecular Formula C18H15ClN4·2HCl
Formula Weight 395.71
Synonym 4-(8-Chloro-2-methyl-11H-imidazo[1, 2-c][2, 3]benzodiazepin-6-benzeneamine dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

309910

 
r & d systems
309910
10mg Each for $252.39

309950

 
r & d systems
309950
50mg Each for $1,050.42
Description & Specifications

Specifications

Chemical Name or Material GYKI 47261 dihydrochloride
CAS 1217049-32-5
Recommended Storage Store at +4°C
Assay Percent Range >99%
Quantity 10mg
Molecular Formula C18H15ClN4·2HCl
Formula Weight 395.71
Synonym 4-(8-Chloro-2-methyl-11H-imidazo[1, 2-c][2, 3]benzodiazepin-6-benzeneamine dihydrochloride
Safety and Handling

Store at +4°C