Please login to your online account to display your discounted pricing

GW 9508, Tocris

Potent and selective FFA1 (GPR40) agonist

$188.60 - $794.96

Chemical Identifiers

Chemical Name or Material GW 9508
Assay Percent Range >99%
Molecular Formula C22H21NO3
Formula Weight 347.41
Synonym 4-[[(3-Phenoxyphenyl)methyl]amino]benzenepropanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

264910

 
r & d systems
RDS264910
10mg Each for $188.60

264950

 
r & d systems
RDS264950
50mg Each for $794.96
Description & Specifications

Specifications

Formula Weight 347.41
Quantity 10mg
CAS 885101-89-3
Assay Percent Range >99%
Molecular Formula C22H21NO3
Synonym 4-[[(3-Phenoxyphenyl)methyl]amino]benzenepropanoic acid
Chemical Name or Material GW 9508
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature