Please login to your online account to display your discounted pricing

GW 791343 hydrochloride, Tocris

P2X7 allosteric modulator

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material GW 791343 hydrochloride
Assay Percent Range >99%
Molecular Formula C20H24F2N4O·3HCl
Formula Weight 483.81
Synonym 2-[(3, 4-Difluorophenyl)amino]-N-[2-methyl-5-(1-piperazinylmethyl)phenyl]-acetamide trihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

338510

 
r & d systems
338510
10mg Each for $239.63

338550

 
r & d systems
338550
50mg Each for $976.15
Description & Specifications

Specifications

Formula Weight 483.81
Quantity 10mg
Molecular Formula C20H24F2N4O·3HCl
Assay Percent Range >99%
Synonym 2-[(3, 4-Difluorophenyl)amino]-N-[2-methyl-5-(1-piperazinylmethyl)phenyl]-acetamide trihydrochloride
Chemical Name or Material GW 791343 hydrochloride
CAS 309712-55-8
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature